Documentos
BAF424004031
Scienceware® Lab Companion cabinet vacuum desiccator
volume 45 L, clear polycarbonate, pkg of 1 ea
Sinónimos:
Desiccator, Desiccator cabinet, Vacuum Bell Jar, Vacuum Desiccator
NACRES:
NB.43
Materiales
clear polycarbonate
envase
pkg of 1 ea
manufacturer/tradename
Bel-Art F42400-4031
apertura de la puerta An × Al
11 5/8 in. × 17 1/2 in.
externo An × L × Al
16 1/2 in. × 15 5/8 in. × 19 3/8 in.
volumen
45 L
45000 mL
¿Está buscando productos similares? Visit Guía de comparación de productos
Comparar elementos similares
Ver comparación completa
Mostrar Diferencias
1 of 4
Este artículo
BAF424004021BAF424004001BAF424004011
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F42400-4031
manufacturer/tradename
Bel-Art F42400-4021
manufacturer/tradename
Bel-Art F42400-4001
manufacturer/tradename
Bel-Art F42400-4011
door opening W × L
11 5/8 in. × 17 1/2 in.
door opening W × L
11 5/8 in. × 9 5/8 in.
door opening W × L
8 1/8 in. × 9 3/8 in.
door opening W × L
11 5/8 in. × 13 5/8 in.
external W × L × H
16 1/2 in. × 15 5/8 in. × 19 3/8 in.
external W × L × H
16 1/2 in. × 15 5/8 in. × 11 in.
external W × L × H
12 5/8 in. × 11 1/4 in. × 10 5/8 in.
external W × L × H
16 1/2 in. × 15 5/8 in. × 15 in.
volume
45 L
volume
23 L , 23000 mL
volume
11 L , 11000 mL
volume
35 L , 35000 mL
Descripción general
These high-quality vacuum desiccators provide vacuum integrity lasting three times longer than the average vacuum desiccator resulting in lower maintenance and greater peace of mind. Able to maintain 29 in.-Hg vacuum for over 72 hours at room temperature, they stay gastight allowing reliable storage or experiments over extended periods.
A Viton® 3-way valve (included) offers consistent and uniform vacuum draw, vacuum release, or gas exchange. Transparent polycarbonate construction is durable, easy-to-clean, chemical-resistant, and allows quick visualization of the desiccator contents. Amber tinted UV blocking models block 100% of UVA, B, and C light to minimize damages or discoloration of light-sensitive samples.
Available in both cabinet and round styles, all models include a desiccant tray (desiccants sold separately).
  • Single piece molded design (except the door) enables outstanding vacuum capability with virtually no leakage and exceptional durability
  • Cabinet style provides maximum useable space
  • Hinged door allows easy access; built in locking tabs secure the door when not under vacuum
  • Analog vacuum gauge in front door allows vacuum level checking at a glance
  • Perforated shelves can be repositioned to accommodate various storage needs (additional shelves sold separately)
  • Built-in ports permit attachment of up to 3 additional valves (sold separately)
  • Stackable space-saving design allows efficient use of work space
  • Eight models to choose from; 11, 23, 35, and 45 L internal volume in clear or UV blocking amber
  • 11 L and 23 L models include two shelves (up to four shelves max.)
  • 35 L and 45 L models include three shelves (up to five and six shelves respectively, max.)
Información legal
Lab Companion is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Certificados de análisis (COA)
¿Ya tiene este producto?
Para su comodidad, los documentos relacionados con los productos que usted ha comprado con anterioridad se han recopilado en la Biblioteca de documentos.
Visite la Librería de documentos
¿Le resulta difícil encontrar su producto o lote/número de lote?
Las referencias de los producto se combinan con los tamaños de envase y las cantidades cuando se muestran en la página web, por ejemplo: T1503-25G. Asegúrese de introducir el número de producto ÚNICAMENTE en el campo del Número de producto (ejemplo: T1503).
Ejemplo
T1503
Referencia del producto
-
25G
Tamaño del envase/cantidad
Otros ejemplos
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
introducir como 1.000309185)
¿Tiene algún problema? No dude en ponerse en contacto con nosotros Servicio técnico para asistencia
Los números de lote se encuentran en la etiqueta del producto después de las palabras «Lot» o «Batch».
Productos Aldrich
  • Si encuentra un número de lote como TO09019TO, introduzca el número de lote 09019TO sin las dos primeras letras.
  • Si encuentra un número de lote con un código de entrada como 05427ES-021, introduzca el número 05427ES sin el código de entrada -021.
  • Si encuentra un número de lote con un código de entrada como STBB0728K9, introduzca el código de lote STBB0728 sin el código de entrada K9.
¿No ha encontrado lo que estaba buscando?
En algunos casos, puede que un COA no esté disponible en la página web. Si su búsqueda no encontró el COA puede solicitarlo.
Solicitar COA