SLW1594/02D
Pyrex® Vacuum desiccator
Metal perforated plate not included; part number SLW1597/02D
Sinónimos:
Pyrex, SciLabware 1594/02D
Seleccione un Tamaño
577,00 €
Seleccione un Tamaño
About This Item
577,00 €
Productos recomendados
Características
Metal perforated plate not included; part number SLW1597/02D
envase
pkg of 1 ea
fabricante / nombre comercial
SciLabware 1594/02D
SciLabware
brida D.I.
220 mm
brida Al total
120 mm , Height from ledge to flange
1 of 4
Este artículo | SLW1591/02D | SLW1593/02D | SLW1590/02D |
---|---|---|---|
packaging pkg of 1 ea | packaging - | packaging pkg of 1 ea | packaging - |
feature Metal perforated plate not included; part number SLW1597/02D | feature Complete desiccators consist of a base and cover. Not supplied with metal perforated plate. | feature - | feature Complete desiccators consist of a base and cover. Not supplied with metal perforated plate |
manufacturer/tradename SciLabware 1594/02D | manufacturer/tradename 1591/02D, SciLabware | manufacturer/tradename 1593/02D, SciLabware | manufacturer/tradename 1590/02D, SciLabware |
flange I.D. 220 mm | flange I.D. 220 mm | flange I.D. 170 mm | flange I.D. 170 mm |
flange Overall H 120 mm , Height from ledge to flange | flange Overall H - | flange Overall H 85 mm , Height from ledge to flange | flange Overall H - |
Descripción general
- Designed for use under vacuum
- Sturdy construction in borosilicate glass
- Ground glass flanges
- Cover has a 24/29 socket to accept desiccator stopcocks
- Plates are positioned on an internal ledge within the base
Información legal
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)