ABE530

Sigma-Aldrich

Anti-Histone-lysine N-methyltransferase SUV39H2 Antibody

from rabbit, purified by affinity chromatography

Sinónimos:
Histone-lysine N-methyltransferase SUV39H2, Histone-lys N-methyltransferase SUV39H2, Histone H3-K9 methyltransferase 2, H3-K9-HMTase 2, Lysine N-methyltransferase 1B, Suppressor of variegation 3-9 homolog 2, Su(var)3-9 homolog 2
eCl@ss:
32160702
NACRES:
NA.41

origen biológico

rabbit

Nivel de calidad

100

antibody product type

primary antibodies

clon

polyclonal

purificado por

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology)

aplicaciones

immunoprecipitation (IP): suitable
western blot: suitable

Nº de acceso NCBI

Nº de acceso UniProt

enviado en

wet ice

Gene Information

human ... SUV39H2(79723)

Descripción general

Histone-lysine N-methyltransferase SUV39H2 is also called Histone H3-K9 methyltransferase 2 (H3-K9-HMTase 2), Lysine N-methyltransferase 1B, and Suppressor of variegation 3-9 homolog 2 (Su(var)3-9 homolog 2). Histone-lysine N-methyltransferase SUV39H2 is a histone methyltransferase that specifically trimethylates ′Lys-9′ of histone H3 using monomethylated H3 ′Lys-9′ as substrate, which creates a specific tag for epigenetic transcriptional repression and directs DNA methylation at pericentric repeats. Histone-lysine N-methyltransferase SUV39H2 is involved in several processes, including cell cycle regulation, transcriptional repression, regulation of telomere length, and higher-order chromatin organization during spermatogenesis.

Inmunógeno

KLH-conjugated linear peptide corresponding to human Histone-lys N-methyltransferase SUV39H2.

Aplicación

Immunoprecipitation Analysis: 10 µg from a representative lot detected Histone-lys N-methyltransferase SUV39H2 in 500 µg of MDA-MB-435 cell lysate.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
Detect SUV39H1 using this rabbit polyclonal antibody, Anti-Histone-lysine N-methyltransferase SUV39H2 Antibody validated for use in western blotting & IP.

Calidad

Evaluated by Western Blotting in human testis tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Histone-lys N-methyltransferase SUV39H2 in 10 µg of human testis tissue lysate.

Descripción de destino

~51 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Forma física

Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Affinity Purfied

Almacenamiento y estabilidad

Stable for 1 year at 2-8°C from date of receipt.

Otras notas

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Cláusula de descargo de responsabilidad

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

WGK Alemania

WGK 1

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable

Certificado de Análisis

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico

Redes sociales

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Investigación. Desarrollo. Producción.

Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos

© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.

Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.