73004BC
Millipore
GDS Adhesive Sheets
BioControl, For use with GDS
manufacturer/tradename
BioControl Systems 73004
condiciones de almacenamiento
room temperature
aplicaciones
PCR: suitable
pathogen testing: suitable
Industria destacada
Food and Beverages
compatibilidad
for use with PICKPEN® PIPETMAX® system
temp. de almacenamiento
room temp
Descripción general
Each sealing film measures 79.4 x 139.7 mm and offers sufficient sealing area for all PCR plates. Length between the perforations with end tabs removed is 125.1 mm.
Componentes
Box of 100 film sheets
Compatibilidad
BioControl GDS
Método de análisis
PCR, Immunomagnetic Separation (IMS)
Información legal
PICKPEN is a registered trademark of BioControl Systems, Inc.
PIPETMAX is a registered trademark of Gilson, Inc
Certificado de Análisis
Certificado de origen