Merck
All Photos(1)
73021BC
Millipore
GDS OTV Insert
BioControl, For use with GDS, Performance verification of the GDS Rotor-Gene
caducidad
3‑5 mo.
manufacturer/tradename
BioControl Systems 73021
condiciones de almacenamiento
protect from light at room temperature
technique(s)
PCR: suitable
application(s)
food and beverages
pathogen testing
compatibilidad
for use with GDS Rotor-Gene
temp. de almacenamiento
room temp
Descripción general
The GDS OTV (Optical Temperature Verification) Insert provides automated performance verification and dynamic calibration of the GDS Rotor-Gene. The OTV system uses the chemical properties of thermochromic liquid crystals (TLC) as an absolute temperature reference. When heated, a TLC changes from opaque to transparent at a very precise temperature. The Rotor-Gene can measure this precise temperature transition point and, by comparing the measured in-tube thermal transition point to the known calibrated value, the OTV software verifies that the Rotor-Gene is within specification. If necessary, the OTV software allows the user to automatically recalibrate the instrument with a simple push of a button. A printable post-run report documents each test and any necessary recalibration.
Componentes
  • OTV Insert – good for 30 verification runs
  • Optical Filters for Rotor-Gene 3000 and Rotor-Gene Q, one each
Compatibilidad
BioControl GDS
Método de análisis
Optical Temperature Verification
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service