73021BC
Millipore
GDS OTV Insert
BioControl, For use with GDS, Performance verification of the GDS Rotor-Gene
caducidad
3‑5 mo.
manufacturer/tradename
BioControl Systems 73021
condiciones de almacenamiento
protect from light at room temperature
aplicaciones
PCR: suitable
pathogen testing: suitable
Industria destacada
Food and Beverages
compatibilidad
for use with GDS Rotor-Gene
temp. de almacenamiento
room temp
Descripción general
The GDS OTV (Optical Temperature Verification) Insert provides automated performance verification and dynamic calibration of the GDS Rotor-Gene. The OTV system uses the chemical properties of thermochromic liquid crystals (TLC) as an absolute temperature reference. When heated, a TLC changes from opaque to transparent at a very precise temperature. The Rotor-Gene can measure this precise temperature transition point and, by comparing the measured in-tube thermal transition point to the known calibrated value, the OTV software verifies that the Rotor-Gene is within specification. If necessary, the OTV software allows the user to automatically recalibrate the instrument with a simple push of a button. A printable post-run report documents each test and any necessary recalibration.
Componentes
  • OTV Insert – good for 30 verification runs
  • Optical Filters for Rotor-Gene 3000 and Rotor-Gene Q, one each
Compatibilidad
BioControl GDS
Método de análisis
Optical Temperature Verification
Certificado de Análisis
Certificado de origen