73026BC
Millipore
GDS Adhesive Strips
BioControl, For use with GDS
manufacturer/tradename
BioControl Systems 73026
condiciones de almacenamiento
room temperature
aplicaciones
PCR: suitable
pathogen testing: suitable
Industria destacada
Food and Beverages
compatibilidad
for use with GDS Rotor-Gene
temp. de almacenamiento
room temp
Descripción general
Each sealing film strip is 19.0 x 97.3 mm, including two removable positioning tabs. The long dimension with tabs removed is 76.2 mm.
Componentes
Box of 200 film strips (50 sheets of four strips)
Compatibilidad
BioControl GDS
Método de análisis
PCR, Immunomagnetic Separation (IMS)
Certificado de Análisis
Certificado de origen