Descripción general
The GDS Vortex Mixer is designed to vortex samples during Assurance® GDS assay sample preparation. Assurance® GDS (Genetic Detection System) is an automated nucleic acid amplification system for the detection of pathogenic organisms in foods, ingredients, and environmental samples. A Vortex mixer is needed for mixing the concentration reagent and sample. It is a variable-speed analog unit, its custom block holder and small orbit are optimal for effective mixing.
NOTE: The vortex mixer is 115V and comes with the US plug only. If located in the EU, please additionally order 73060BC for the European power converter and plug.
Aplicación
The GDS Vortex Mixer is designed to vortex the concentration reagent and samples during Assurance® GDS assay sample preparation.
Componentes
- Vortex Mixer
- Microtiter plate attachment
Compatibilidad
BioControl GDS
Método de análisis
PCR, Immunomagnetic Separation (IMS)
Información legal
Assurance is a registered trademark of BioControl Systems, Inc.
Assurance GDS is a registered trademark of BioControl Systems, Inc.