Merck
All Photos(3)
73070BC
Millipore
GDS Rotor-Gene Q Thermocycler
Advanced rotary thermocycler for use with Assurance® GDS PCR assays
manufacturer/tradename
Assurance GDS®
BioControl Systems 73070
technique(s)
PCR: suitable
application(s)
food and beverages
pathogen testing
temp. de almacenamiento
room temp
Descripción general
Assurance® GDS (Genetic Detection System) is an automated nucleic acid amplification system for the detection of pathogenic organisms in foods, ingredients, and environmental samples. The unique centrifugal rotary design of the Rotor-Gene® Q makes it the most precise and versatile thermocycler currently available. Each tube spins in a chamber of moving air, keeping all samples at precisely the same temperature during rapid thermal cycling thereby enabling uniform detection. When each tube aligns with the detection optics, the sample is illuminated, and the fluorescent signal is rapidly collected from a single, short optical pathway. This thermal and optical uniformity results in sensitive, precise, and fast PCR analysis. It also eliminates sample-to-sample variations and edge effects. These are unavoidable in traditional block-based instruments due to temperature gradients across the block and multiple, complex optical pathways.
Aplicación
GDS Rotor-Gene® Q Thermocycler is used with Assurance® GDS assays for the detection of pathogenic organisms in foods, ingredients, and environmental samples without the need for optimization of reaction and cycling conditions.
Envase
1 each
Características y beneficios
The rotary design delivers:
  • Tube-to-tube variation ±0.02°C for uniform detection
  • Pre-loaded firmware specific to Assurance® GDS assays
  • Negligible equilibration times for fast run-cycling
  • Complete confidence in your results
Componentes
  • GDS Rotor-Gene Q
  • User Manual
  • GDS Gel Cooling Block (pack of 2)
Compatibilidad
BioControl GDS
Método de análisis
PCR
Información legal
Assurance is a registered trademark of BioControl Systems, Inc.
Assurance GDS is a registered trademark of BioControl Systems, Inc.
Rotor-Gene is a registered trademark of Qiagen GmbH
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service