Merck
All Photos(1)

73071BC

Millipore

GDS Rotor-Gene Q Laptop

Laptop for use with Assurance® GDS Rotor-Gene® Q thermocycler

manufacturer/tradename

Assurance GDS®
BioControl Systems 73071

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibilidad

for use with GDS Rotor-Gene

temp. de almacenamiento

room temp

Descripción general

GDS Rotor-Gene® Q thermocycler is supported with Dell Latitude laptop. Dell Latitude laptops work faster and provides flexible, simplified and secure solutions. The laptop is made with premium finishes like carbon fiber and all new aluminum, ensuring reimagined design and durability in a lighter frame. The laptop is pre-installed with the comprehensive Rotor-Gene® Q software package that supports the state-of-the art real-time analysis procedures from basic to advanced algorithms. The software determines sample results for the user and increases the reliability of results. Data security is assured, and all process steps are trackable from starting the run to exporting the results.

Aplicación

GDS Rotor-Gene® Q Laptop pre-installed with the software determines sample results for the user and increases the reliability of results with assured data security.

Envase

1 each

Características y beneficios

  • BioControl GDS software pre-installed
  • Optimally configured for Assurance® GDS
  • 3 User profiles:
1. The “GDS Administrator” user level provides administrators with the ability to set up new users, Password protected, updated software, and configure the computer′s operational settings.
2. The “GDS Analyst” user level provides laboratory managers with the ability to analyze data in greater detail. Analysts have the ability to generate reports that can be saved, printed, or e-mailed directly from the GDS Rotor-Gene® software and is password protected.
3. The “GDS Operator” is the default access level that allows the user to enter sample information and provides a convenient results table.

Componentes

  • Dell Latitude laptop
  • A/C Adapter

Compatibilidad

BioControl GDS

Método de análisis

PCR

Información legal

Assurance is a registered trademark of BioControl Systems, Inc.
Assurance GDS is a registered trademark of BioControl Systems, Inc.
Rotor-Gene is a registered trademark of Qiagen GmbH

Código de clase de almacenamiento

11 - Combustible Solids

WGK

WGK 3

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service