Merck
All Photos(1)

73076BC

Millipore

GDS 72 Well Rotor, RG Q Only

BioControl Systems, for use with GDS Rotor-Gene Thermocycler

Synonym(s):
Thermocycler Rotor

manufacturer/tradename

BioControl Systems

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibilidad

for use with GDS Rotor-Gene Thermocycler

temp. de almacenamiento

room temp

Descripción general

GDS 72 Well Rotor is an accessory to hold the amplification tubes (0.1 mL) containing samples in the GDS Rotor-Gene® Q thermocycler during amplification. GDS is an automated nucleic acid amplification system for the detection of pathogenic organisms in foods, ingredients, and environmental samples. The unique centrifugal rotary design of the Rotor-Gene® Q makes it the most precise and versatile thermocycler. Amplification tubes spin in a chamber of moving air, keeping all samples at precisely the same temperature during rapid thermal cycling thereby enabling uniform detection. When each tube aligns with the detection optics, the sample is illuminated, and the fluorescent signal is rapidly collected from a single, short optical pathway. This thermal and optical uniformity results in sensitive, precise, and fast PCR analysis.

Aplicación

GDS 72 well Rotor is used with Assurance® GDS Rotor-Gene® Q thermocycler for the detection of pathogenic organisms in foods, ingredients, and environmental samples without the need for optimization of reaction and cycling conditions.

Envase

1 each

Características y beneficios

The rotor design delivers:
  • Tube-to-tube variation of ±0.02°C for uniform detection
  • Pre-loaded firmware specific to Assurance® GDS assays
  • Negligible equilibration times for fast run-cycling
  • Complete confidence in your results

Información legal

Assurance is a registered trademark of BioControl Systems, Inc.
Rotor-Gene is a registered trademark of Qiagen GmbH

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service