Merck
All Photos(1)

73087BC

Millipore

Assurance® GDS PickPen® Tips

BioControl, For use with GDS PickPen®

Synonym(s):
PickPen® Tips

envase

rack of 96 tips

manufacturer/tradename

BioControl Systems 73087

condiciones de almacenamiento

room temperature

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibilidad

for use with PICKPEN®

temp. de almacenamiento

room temp

Descripción general

GDS PickPen® Tips are the consumable accessories of the GDS PickPen® device. Assurance® GDS is an automated nucleic acid amplification system for the detection of pathogenic organisms in foods, ingredients, and environmental samples. As part of the sample preparation, the proprietary GDS PickPen® is designed to transfer magnetic particles when working with Assurance® GDS pathogen detection kits. The incorporation of the GDS PickPen® in the immunomagnetic separation (IMS) process improves the procedure by concentrating the target pathogen while decreasing the carryover of background microflora from a wide range of food matrices without compromising the sensitivity.

Aplicación

The Assurance® GDS PickPen® Tips are used with the PickPen® device to collect and transfer magnetic particles during sample preparation for detection of food pathogens by Assurance® GDS.

Componentes

One rack of 96 PickPen tips

Compatibilidad

BioControl GDS

Método de análisis

Immunomagnetic Separation (IMS)

Información legal

Assurance is a registered trademark of BioControl Systems, Inc.
PICKPEN is a registered trademark of BioControl Systems, Inc.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service