Merck
All Photos(2)
73091BC
Millipore
GDS PickPen®, 8-Magnets
Immunomagnetic separation device for use with Assurance® GDS PCR system
manufacturer/tradename
Assurance GDS®
BioControl Systems 73091
technique(s)
PCR: suitable
application(s)
food and beverages
pathogen testing
compatibilidad
for use with GDS Rotor-Gene
temp. de almacenamiento
room temp
Descripción general
Assurance® GDS (Genetic Detection System) is an automated nucleic acid amplification system for the detection of pathogenic organisms in foods, ingredients, and environmental samples. The proprietary GDS PickPen®, 8-Magnets is designed to transfer magnetic particles when working with Assurance® GDS pathogen detection kits. The incorporation of the PickPen® in the immunomagnetic separation (IMS) process improves the procedure by concentrating the target pathogenwhile decreasing carryover of background microflora without compromising the sensitivity.
Aplicación
The GDS PickPen®, 8-Magnets is designed to transfer magnetic particles when working with Assurance® GDS pathogen detection kits used for the detection of pathogenic organisms in foods, ingredients, and environmental samples.
Envase
1 each
Características y beneficios
  • Compatible with all Assurance® GDS disposable wells/plates, 8 samples can be taken through IMS at one time
  • Consistent recovery of immuno-magnetic particles during IMS
  • Lower carryover of background microflora
Componentes
One GDS PickPen
Compatibilidad
BioControl GDS
Método de análisis
Immunomagnetic Separation (IMS)
Información legal
Assurance is a registered trademark of BioControl Systems, Inc.
Assurance GDS is a registered trademark of BioControl Systems, Inc.
PICKPEN is a registered trademark of BioControl Systems, Inc.
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service