Merck
All Photos(1)
79174BC
Millipore
GDS PPMX Gel Cooling Block
BioControl, For use with GDS PickPen® PIPETMAX®
material
plastic rack
manufacturer/tradename
BioControl Systems 79174
condiciones de almacenamiento
room temperature
technique(s)
PCR: suitable
application(s)
food and beverages
pathogen testing
compatibilidad
for use with GDS PICKPEN® PIPETMAX® System
temp. de almacenamiento
room temp
Descripción general
4°C, Pink/Purple, 96 Position Rack
Envase
1 each
Características y beneficios
  • Maintain a sample temperature of less than 4°C for almost four hours (with lid on rack) at room temperature.
  • The plastic changes from purple to pink as the temperature of the rack changes, enabling you to easily monitor the temperature of your samples.
Componentes
One box of 2 cooling blocks for PPMX
Compatibilidad
BioControl GDS
Método de análisis
PCR
Información legal
PICKPEN is a registered trademark of BioControl Systems, Inc.
PIPETMAX is a registered trademark of Gilson, Inc
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service