Merck
All Photos(1)

PE1126

Supelco

Color - WP

Proficiency Testing Material, Concentrate for dilution to 100 mL

Synonym(s):
CWA - Color, WP Color
NACRES:
NA.24

Nivel de calidad

envase

ampule of 2 × 20 mL

concentración

in water

application(s)

environmental

formato

matrix material

Descripción general

Color - Water Pollution (WP) proficiency testing material is produced in accordance with ISO/IEC 17043:2010. Also referred to as non-Potable water or Water Pollution sample, this product is produced according to TNI requirements. However, please note, this sample may include non-TNI analytes as well. The higher criteria from the TNI tables mimic the levels common to wastewater samples. The PT portfolio allows our test components to offer significant performance data to monitor and prove competency to customers, accreditation bodies or both.

Supelco® is a global proficiency testing (PT) provider that manufactures best-in-class environmental and pharmaceutical PT products. Most water PT samples are available as concentrates or ready-to-use volumes giving the end-user flexibility in selecting samples that will fit their exact needs.
These samples are typically provided in duplicate for use in case of lab mishap, for QC purposes or to analyze multiple methods. 

Type of PT Scheme: Water analysis - Water Pollution (WP)

Matrix: Water Pollution/Wastewater

Analyte/s: Values of analytes vary from lot to lot.

All information regarding the use of Color - Water Pollution (WP) Proficiency Testing standard can be found in the reporting packet which will be available to download at PT Portal.

Aplicación

Color - Water Pollution (WP) proficiency testing material is used for environmental analysis particularly in Water Pollution/Wastewater samples.

Have any questions? Explore our FAQs.
For more information on this product, please contact our PT Service Team.

Envase

20 mL

Características y beneficios

We have a vast portfolio of environmental PT schemes with "fit-for-purpose" samples to help you meet the needs of the laboratory.

Our proficiency testing (PT) program is a benchmark to:

  • provide staff with an insight into their laboratory′s performance, to verify the accuracy and reliability of their testing process
  • provide a comparison of performance with that of other laboratories, nationally and internationally
  • test and educate staff, provide a better understanding of the impact of incorrect results and identify areas where there may be problems
  • provide an independent evaluation of laboratory performance
  • demonstrate to clients, colleagues and accreditation bodies that there is a commitment to quality

Nota de preparación

Description

  • This PT sample is a 20 mL concentrate.
  • The solvent for this sample is water.
  • The PT sample has been autoclaved for preservation.

Sample Preparation

  • To prepare the solution, partially fill a 100 mL volumetric flask with reagent-grade water (near 20 ºC).
  • Open ampoule PE1126-20ML (be careful to avoid cuts on glass) and transfer 10.0 mL of concentrate to the volumetric flask.
  • Fill the flask to volume with reagent-grade water and mix well.
  • The sample is now ready for immediate analysis.
  • Report only the properties for the method you are seeking accreditation.

Almacenamiento y estabilidad

Please refer to sample label for expiration date, specific storage temperature, and specific storage conditions.

Otras notas

Are you new to Supelco® Proficiency Testing? Be sure to register your account details in the PT Portal before placing your order.
This will provide access to data entry, reporting packets and final reports. Registrations will be approved shortly after your order is placed. For further questions, write to us at ptservice@milliporesigma.com.

When placing your order online, please indicate your study preference under the ′your reference′ column. Available studies can be viewed at the bottom of the page.
Click here to learn more about Quick-Turn Studies (on-demand proficiency tests that are offered depending on lot availability). The Quick-Turn results are released within 48-72 hours whereas scheduled study reports are released within 21 days.

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes

Proficiency Testing Schedule (2022-2023)


Wastewater
WP22-1 January 12 – February 25, 2022
WP22-2 March 23 - May 6, 2022
WP22-3 July 6 - August 19, 2022
WP22-4 September 21 - November 4, 2022

WP23-1 January 11 – February 24, 2023
WP23-2 March 22 – May 5, 2023
WP23-3 July 5 – August 18, 2023
WP23-4 September 20 – November 3, 2023


Quick-Turn Study (on demand proficiency test that is offereddepending on lot availability)

Información legal

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Color 10.0 - 75.0

pictogramas

Health hazardEnvironment

Palabra de señalización

Danger

Frases de peligro

Clasificaciones de peligro

Aquatic Chronic 2 - Carc. 1B

Código de clase de almacenamiento

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service