103A-7

Sigma-Aldrich

CD3 Rabbit Polyclonal Antibody

NACRES:
NA.41

origen biológico

rabbit

Nivel de calidad

500
100

forma del anticuerpo

Ig fraction of antiserum

antibody product type

primary antibodies

clon

polyclonal

descripción

For In Vitro Diagnostic Use in Select Regions (See Chart)

formulario

buffered aqueous solution

species reactivity

human

envase

vial of 0.5 mL concentrate (103A-75)
bottle of 1.0 mL predilute (103A-77)
vial of 1.0 mL concentrate (103A-76)
bottle of 7.0 mL predilute (103A-78)

manufacturer/tradename

Cell Marque

aplicaciones

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:1000

conjugado

unconjugated

control

tonsil

enviado en

wet ice

temp. de almacenamiento

2-8°C

visualización

membranous

Categorías relacionadas

Descripción general

Anti-CD3 antibody has been considered the best all around T-cell marker. This antibody reacts with an antigen present in early thymocytes. The positive staining of this marker may represent a sign of early commitment to the T-cell lineage.

Ligadura / enlace

CD3 Positive Control Slides, Product No. 103S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Forma física

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Otras notas

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Información legal

Cell Marque is a trademark of Sigma-Aldrich Co. LLC

RIDADR

NONH for all modes of transport

Certificado de Análisis
SM Denning, et al.
Leucocyte Typing III, 144-147 (1987)
Kennosuke Karube et al.
The American journal of surgical pathology, 27(10), 1366-1374 (2003-09-26)
We studied the morphologic, immunohistochemical, and clinical characteristics of 158 cases of lymphoblastic lymphoma. Based on immunophenotyping and cell lineage, cases were classified into B-cell type (CD20,CD19 or CD79a+, n = 53), T-cell type (surface CD3+, n = 84), and...
E A Clark et al.
Immunology today, 10(7), 225-228 (1989-07-01)
During 1987, striking advances were made in defining the receptors and ligands for cell-to-cell adhesion interactions involving leukocytes. In 1988, two major leukocyte differentiation antigens, CD10 (cALLA) and CD45 (LCA, T200), were shown to be enzymes while two other markers...
Ulla Axdorph et al.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica, 110(5), 379-390 (2002-06-22)
Morphologically, T-cell-rich B-cell lymphoma (TCRB-NHL) may be indistinguishable from Hodgkin's disease (HD). Immunophenotyping may be helpful in the separation of these entities. TCRB-NHL is occasionally misdiagnosed and treated as HD. However, information is limited regarding clinical characteristics and outcome of...
Ahmet Dogan et al.
The American journal of surgical pathology, 27(7), 903-911 (2003-06-27)
Occasionally, primary large B-cell lymphomas (LBLs) arising in the spleen present with a micronodular pattern involving the splenic white pulp but sparing the red pulp. Histologically, the nodules contain scattered large B cells in a background of numerous T cells...

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico

Redes sociales

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Investigación. Desarrollo. Producción.

Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos

© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.

Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.