Saltar al contenido
Merck

AMAB91795

Anti-TMPRSS2 antibody produced in mouse

Prestige Antibodies® Powered by Atlas Antibodies, clone CL12514, purified by using Protein A, buffered aqueous glycerol solution

Sinónimos:

PRSS10


Iniciar sesión para ver los precios por organización y contrato.

Seleccione un Tamaño

En este momento no podemos mostrarle ni los precios ni la disponibilidad

Acerca de este artículo

NACRES:
NA.41
UNSPSC Code:
12352203

Saltar a

Servicio técnico
¿Necesita ayuda? Nuestro equipo de científicos experimentados está aquí para ayudarle.
Permítanos ayudarle

Nombre del producto

Anti-TMPRSS2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL12514, purified by using Protein A, buffered aqueous glycerol solution

biological source

mouse

conjugate

unconjugated

antibody product type

primary antibodies

clone

CL12514, monoclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

purified by

using Protein A

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:1000-1:2500

isotype

IgG2a

Ensembl | human accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Quality Level

Gene Information

human ... TMPRSS2(7113)

Comparar elementos similares

Ver comparación completa

Mostrar Diferencias

1 of 4

Este artículo
AMAB91793SAB1400226HPA072934
biological source

mouse

biological source

mouse

biological source

mouse

biological source

rabbit

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

clone

CL12514, monoclonal

clone

CL12512, monoclonal

clone

polyclonal

clone

polyclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Gene Information

human ... TMPRSS2(7113)

Gene Information

human ... TMPRSS2(7113)

Gene Information

human ... PRSS2(5645)

Gene Information

human ... TMPRSS15(5651)

shipped in

wet ice

shipped in

wet ice

shipped in

dry ice

shipped in

wet ice

Application

All Prestige Antibodies® Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

All Prestige Antibodies® Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Immunogen

Transmembrane serine protease 2

Physical form

The antibodies are delivered in 40% glycerol and PBS (pH 7.2). 0.02% sodium azide is added as preservative.

Preparation Note

Store at +4°C for short term storage. Long time storage is recommended at -20°C.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

¿No encuentra el producto adecuado?  

Pruebe nuestro Herramienta de selección de productos.

Clase de almacenamiento

10 - Combustible liquids

wgk

WGK 1


Elija entre una de las versiones más recientes:

Certificados de análisis (COA)

Lot/Batch Number

¿No ve la versión correcta?

Si necesita una versión concreta, puede buscar un certificado específico por el número de lote.

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos

Preguntas

Revisiones

Sin puntuación

Filtros activos

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico