SAB1412255

Sigma-Aldrich

ANTI-FOXM1 antibody produced in mouse

clone 2E2, purified immunoglobulin, buffered aqueous solution

Sinónimos:
HFH-11, FOXM1, HFH11, FKHL16, FOXM1B
NACRES:
NA.41

Nivel de calidad

100

origen biológico

mouse

forma del anticuerpo

purified immunoglobulin

antibody product type

primary antibodies

clon

2E2, monoclonal

formulario

buffered aqueous solution

mol peso

antigen 35.53 kDa

species reactivity

human

aplicaciones

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotipo

IgG2aκ

conjugado

unconjugated

Nº de acceso NCBI

Nº de acceso UniProt

enviado en

dry ice

temp. de almacenamiento

−20°C

Gene Information

human ... FOXM1(2305)

Inmunógeno

FOXM1 (NP_973731, 22 a.a. ~ 110 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
NAPSETSEEEPKRSPAQQESNQAEASKEVAESNSCKFPAGIKIINHPTMPNTQVVAIPNNANIHSIITALTAKGKESGSSGPNKFILIS

Forma física

Solution in phosphate buffered saline, pH 7.4

Cláusula de descargo de responsabilidad

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

storage_class_code

12 - Non Combustible Liquids

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable

Certificado de Análisis

Certificado de origen

Xiaoxiao Li et al.
Clinical and experimental medicine, 14(4), 447-455 (2013-10-09)
The transcription factor forkhead box protein M1 (FoxM1) and tyrosine kinase receptor Her-2, aberrantly expressing in various kinds of human malignancies, are closely related to the development of cancer. However, the correlation of them is still little explored, especially in...
Guoyan Liu et al.
The Journal of pathology, 233(3), 308-318 (2014-03-08)
Ovarian carcinoma is the most lethal gynaecological malignancy. Better understanding of the molecular pathogenesis of this disease and effective targeted therapies are needed to improve patient outcomes. MicroRNAs play important roles in cancer progression and have the potential for use...
Satoru Inoguchi et al.
FEBS letters, 588(17), 3170-3179 (2014-07-08)
Here, we found that microRNA-24-1 (miR-24-1) is significantly reduced in bladder cancer (BC) tissues, suggesting that it functions as a tumour suppressor. Restoration of mature miR-24-1 inhibits cancer cell proliferation and induces apoptosis. Forkhead box protein M1 (FOXM1) is a...

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico