Merck
All Photos(1)

SAB4301577

Sigma-Aldrich

Anti-phospho-GATA3 (pSer308) antibody produced in rabbit

affinity isolated antibody

Synonym(s):
HDRS, HDR
NACRES:
NA.41

origen biológico

rabbit

Nivel de calidad

100

forma del anticuerpo

affinity isolated antibody

antibody product type

primary antibodies

clon

polyclonal

formulario

buffered aqueous solution

mol peso

47 kDa

species reactivity

mouse, rat, human

concentración

1.0 mg/mL

technique(s)

western blot: 1:500-1:1000 (Cell Lysate)

isotipo

IgG

nº de acceso

NP_001002295.1

Nº de acceso UniProt

enviado en

wet ice

temp. de almacenamiento

−20°C

Gene Information

human ... GATA3(2625)

Especificidad

The antibody detects endogenous level of GATA3 only when phosphorylated at Serine 308.

Inmunógeno

Peptide sequence around phosphorylation site of Serine 308(R-L-S(p)-A-A) derived from Human GATA3.

Características y beneficios

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Forma física

Supplied at 1.0mg/mL in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Cláusula de descargo de responsabilidad

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Código de clase de almacenamiento

10 - Combustible liquids

WGK

WGK 1

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service