Merck
All Photos(1)

SAB4700739

Sigma-Aldrich

Monoclonal Anti-CD28 antibody produced in mouse

clone JJ319, purified immunoglobulin, buffered aqueous solution

NACRES:
NA.41

Nivel de calidad

100

origen biológico

mouse

conjugado

unconjugated

forma del anticuerpo

purified immunoglobulin

antibody product type

primary antibodies

clon

JJ319, monoclonal

formulario

buffered aqueous solution

species reactivity

rat

concentración

1 mg/mL

technique(s)

FUNC: suitable
flow cytometry: suitable
immunoprecipitation (IP): suitable

isotipo

IgG1

Nº de acceso NCBI

Nº de acceso UniProt

enviado en

wet ice

temp. de almacenamiento

2-8°C

Gene Information

rat ... Cd28(25660)

Descripción general

The mouse monoclonal antibody JJ319 reacts with CD28, a disulfide-linked homodimeric type I glycoprotein (monomer of Mw 44 kDa) which is a critical costimulatory receptor of T cells.

Inmunógeno

Mouse A20J B lymphoma cells transfected with rat CD28

Características y beneficios

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Forma física

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Cláusula de descargo de responsabilidad

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Código de clase de almacenamiento

12 - Non Combustible Liquids

Punto de inflamabilidad F

Not applicable

Punto de inflamabilidad C

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service