821400
TSKgel® Hardware and Accessories
polycarbonate housing, polypropylene cap, pkg of 1 ea
Sinónimos:
ToyoScreen Column Holder
material
polycarbonate housing
polypropylene cap
envase
pkg of 1 ea
manufacturer/tradename
Tosoh 21400
¿Está buscando productos similares? Visit Guía de comparación de productos
Descripción general
Every large scale commercial purification begins at a small scale in a method development lab by evaluating retention and selectivity of the target protein versus impurities, host cell proteins, DNA, etc., on multiple resins, often from different vendors. Depending on the constraints, the process development engineer can either purchase bulk resins to pack several small columns with different resins or take advantage of the speed and convenience of pre-packed columns for resin evaluation. Pre-packed ToyoScreen® Process Development Columns are available in 1 mL and 5 mL volumes packed with the most popular Toyopearl® resins. ToyoScreen columns can be used to quickly establish optimum elution conditions. Varying pH, salt type, gradient conditions and flow rate are common experimental parameters explored. Initial results from resin screening and optimization with ToyoScreen columns accurately predict the separation behavior at larger scale.
Productos recomendados
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Información legal
TSKgel is a registered trademark of Tosoh Corporation
ToyoScreen is a registered trademark of Tosoh Corporation
Toyopearl is a registered trademark of Tosoh Corporation