Merck
  • ES5263
All Photos(1)
ES5263
GreenSep HILIC SFC 5 Micron HPLC Column
5 μm particle size, L × I.D. 15 cm × 3.0 mm
NACRES:
SB.52
material
stainless steel column
Nivel de calidad
Línea del producto
GreenSep
manufacturer/tradename
ES Industries 133291-GS-HILIC
technique(s)
HPLC: suitable
L × D.I.
15 cm × 3.0 mm
matriz
fully porous particle
grupo activo de la matriz
HILIC phase
tamaño de partícula
5 μm
tamaño de poro
120 Å pore size
operating pH
2-8
técnica de separación
supercritical fluid (SFC)
Looking for similar products? Visit Product Comparison Guide
Descripción general
GreenSep HILIC is composed of a polyhydroxylated polymer that is coated and bound to silica. This composition provides hydroxyl levels, that are well above conventional silica and diol type stationary phases, and permits higher loading. The chromatogram below is an example of the superior separation performance obtainable for chemicals containing a variety of functional groups with the GreenSep HILIC column.
Productos recomendados
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Información legal
GreenSep is a trademark of ES Industries
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service