ES5263
stainless steel column
GreenSep™
ES Industries 133291-GS-HILIC
HPLC: suitable
15 cm × 3.0 mm
fully porous particle
HILIC phase
5 μm
120 Å pore size
2-8
supercritical fluid (SFC)
Looking for similar products? Visit Product Comparison Guide
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service