ES5355
GreenSep Pyridyl Amide SFC 5 Micron HPLC Column
5 μm particle size, L × I.D. 15 cm × 3.0 mm
NACRES:
SB.52
manufacturer/tradename
ES Industries 133291-GS-PYA
Nivel de calidad
100
aplicaciones
HPLC: suitable
L × D.I.
15 cm × 3.0 mm
grupo activo de la matriz
pyridyl amide phase
tamaño de partícula
5 μm
tamaño de poro
120 Å pore size
técnica de separación
supercritical fluid (SFC)
¿Está buscando productos similares? Visit Guía de comparación de productos
Descripción general
A metal free, ultra high purity chromatographic media that is pressure stable and specifically engineered for high performance SFC separations. The surface is treated to produce optimum SFC separation interactions and loading capacity while maintaining superior peak shape perfor- mance for many chemicals. GreenSep Silica is available for analytical and preparative column formats in particle sizes from 1.8 μm to 20 μm.
Productos recomendados
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Información legal
GreenSep is a trademark of ES Industries