Nombre del producto
Activated charcoal powder
Referencia del producto
05105
Marca de producto
SIAL
Número de CAS
7440-44-0
Peso molecular
12.01
Ensayo
Especificación
APPEARANCE (COLOR)Black
APPEARANCE (FORM)Powder
RESIDUE ON IGNITION≤ 2.0 %
APPLICATION TESTADSORPTION OF METHYLENE BLUE AND IODINE: CORRESPONDS
METAL TRACE ANALYSIS (ICP)CORRESPONDS TO REQUIREMENTS
CALCIUM (ICP)≤ 1000 mg/kg
CADMIUM (ICP)≤ 10 mg/kg
COBALT (ICP)≤ 10 mg/kg
CHROMIUM (ICP)≤ 10 mg/kg
COPPER (ICP)≤ 10 mg/kg
IRON (ICP)≤ 300 mg/kg
POTASSIUM (ICP)≤ 200 mg/kg
MAGNESIUM (ICP)≤ 200 mg/kg
MANGANESE (ICP)≤ 50 mg/kg
SODIUM (ICP)≤ 500 mg/kg
NICKEL (ICP)≤ 10 mg/kg
LEAD (ICP)≤ 10 mg/kg
ZINC (ICP)≤ 10 mg/kg
TOTAL SULFUR AS SO4 (ICP)≤ 5000 mg/kg
CHLORIDE (CL)≤ 5000 mg/kg
RECOMMENDED RETEST PERIOD72 MONTHS