Nombre del producto
Anti-Interleukin-4 antibody produced in goat affinity isolated antibody, lyophilized powder
Referencia del producto
I7526
Marca de producto
SIGMA
Número de CAS
Peso molecular
Ensayo
Especificación
Bioactivity0.005 - 0.250 µg/ml
 the typical value for ND50 is 0.02-0.1 ug/mL IL-4
 Neutralization, TF-1 Cells) Results outside this
 range are acceptable based on concurrent, side by
 side comparisons with a previously passed lot as
 control.
Titer by Direct ELISAConforms
 0.5-1.0 ug/ml detects 1 ng/well of recombinant
 human IL-4
Working DilutionConforms
 by Indirect Immunoblotting 0.1-0.2 ug/mL detects 2
 ng IL-4/Lane of under reducing and non-reducing
 conditions
Endotoxin Assay≤0.1
 EU/uG antibody
FormulationConforms
 Lyophilized from 0.2 uM Filtered solution in
 phosphate buffered saline with 5% Trehalose