Merck
Dimethyl Sulfoxide - CAS 67-68-5 - Calbiochem ACS Reagent Grade.

Dimethyl Sulfoxide - CAS 67-68-5 - Calbiochem

Dimethyl Sulfoxide - CAS 67-68-5 - Calbiochem ACS Reagent Grade.
Synonyms:
Dimethyl sulfoxide, DMSO
Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Dimethyl Sulfoxide - CAS 67-68-5 - Calbiochem, ACS Reagent Grade.