ES
ES
Aplicaciones
Productos
Servicios
Asistencia técnica
Home
Search Results
pH - WS
(1)
pH - WS
Synonyms:
pH-WS
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
PE1368
pH - WS, Proficiency Testing Material
Expand
View Pricing