Merck
Potassium dihydrogen phosphate anhydrous for HPLC LiChropur™

Potassium dihydrogen phosphate anhydrous

Potassium dihydrogen phosphate anhydrous for HPLC LiChropur™
Synonyms:
Potassium phosphate monobasic, Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate
Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Potassium dihydrogen phosphate anhydrous, for HPLC LiChropur