ES
ES
Aplicaciones
Productos
Servicios
Asistencia técnica
Home
Search Results
Sodium bicarbonate solution
(1)
Sodium bicarbonate solution
CAS Number:
144-55-8
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
S8761
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Expand
View Pricing