ES
ES
Aplicaciones
Productos
Servicios
Asistencia técnica
Home
Search Results
Maternal and neonatal urinary excretion of sulfate and glucuronide ritodrine conjugates
Maternal and neonatal urinary excretion of sulfate and glucuronide ritodrine conjugates
Clinical Pharmacology and Therapeutics (1988)
Brashear WT, et al.