• Inicio
  • Resultados de la búsqueda
  • Identification of miRNomes in human liver and hepatocellular carcinoma reveals miR-199a/b-3p as therapeutic target for hepatocellular carcinoma.
Identification of miRNomes in human liver and hepatocellular carcinoma reveals miR-199a/b-3p as therapeutic target for hepatocellular carcinoma.
Cancer cell (2011-02-15)
Jin Hou, Li Lin, Weiping Zhou, Zhengxin Wang, Guoshan Ding, Qiongzhu Dong, Lunxiu Qin, Xiaobing Wu, Yuanyuan Zheng, Yun Yang, Wei Tian, Qian Zhang, Chunmei Wang, Qinghua Zhang, Shi-Mei Zhuang, Limin Zheng, Anmin Liang, Wenzhao Tao, Xuetao Cao
RESUMEN
The full scale of human miRNome in specific cell or tissue, especially in cancers, remains to be determined. An in-depth analysis of miRNomes in human normal liver, hepatitis liver, and hepatocellular carcinoma (HCC) was carried out in this study. We found nine miRNAs accounted for ∼88.2% of the miRNome in human liver. The third most highly expressed miR-199a/b-3p is consistently decreased in HCC, and its decrement significantly correlates with poor survival of HCC patients. Moreover, miR-199a/b-3p can target tumor-promoting PAK4 to suppress HCC growth through inhibiting PAK4/Raf/MEK/ERK pathway both in vitro and in vivo. Our study provides miRNomes of human liver and HCC and contributes to better understanding of the important deregulated miRNAs in HCC and liver diseases.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys9) Antibody, clone 6F12-H4, clone 6F12-H4, from mouse
Sigma-Aldrich
Acetyl-Histone H4 Immunoprecipitation (ChIP) Assay Kit, Acetyl-Histone H4 Immunoprecipitation (ChIP) Assay Kit used to immunoprecipitate transcriptionally active chromatin from mammalian cells using anti-Acetyl-Histone H4, ChIP grade rabbit antiserum.
Redes sociales
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Investigación. Desarrollo. Producción.
Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos
© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.
Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.