• Inicio
  • Resultados de la búsqueda
  • Changes in bacterial diversity and catabolic gene abundance during the removal of dimethylphenol isomers in laboratory-scale constructed wetlands.
Changes in bacterial diversity and catabolic gene abundance during the removal of dimethylphenol isomers in laboratory-scale constructed wetlands.
Applied microbiology and biotechnology (2018-11-12)
Mónica A Vásquez Piñeros, Paula M Martínez-Lavanchy, Kristin Schmidt, Melina Mardones, Hermann J Heipieper
RESUMEN
Constructed wetlands (CWs) are well-established wastewater treatment technologies and applied for bioremediation of contaminated water. Despite the optimal performance of CWs, the understanding of the bacterial processes in the rhizosphere, where mainly microbial degradation processes take place, is still limited. In the present study, laboratory-scale CWs planted with Juncus effusus and running under controlled conditions were studied in order to evaluate removal efficiency of dimethylphenols (DMPs), also in comparison to an unplanted bed. Next to removal rates, the bacterial community structure, diversity, and distribution, their correlation with physiochemical parameters, and abundance of the phenol hydroxylase gene were determined. As a result, better removal performance of DMP isomers (3,4-, 3,5-, and 2,6-DMP added as singles compounds or in mixtures) and ammonium loads, together with a higher diversity index, bacterial number, and phenol hydroxylase gene abundance in Juncus effusus CW in comparison with the non-planted CW, indicates a clear rhizosphere effect in the experimental CWs. An enhancement in the DMP removal and the recovery of the phenol hydroxylase gene were found during the fed with the DMP mixture. In addition, the shift of bacterial community in CWs was found to be DMP isomer dependent. Positive correlations were found between the bacteria harboring the phenol hydroxylase gene and communities present with 3,4-DMP and 3,5-DMP isomers, but not with the community developed with 2,6-DMP. These results indicate that CWs are highly dynamic ecosystems with rapid changes in bacterial communities harboring functional catabolic genes.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Glass beads, acid-washed, ≤106 μm (−140 U.S. sieve)
Redes sociales
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Investigación. Desarrollo. Producción.
Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos
© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.
Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.