  • Search Results
  • The MYC Oncogene Cooperates with Sterol-Regulated Element-Binding Protein to Regulate Lipogenesis Essential for Neoplastic Growth.
The MYC Oncogene Cooperates with Sterol-Regulated Element-Binding Protein to Regulate Lipogenesis Essential for Neoplastic Growth.
Cell metabolism (2019-08-27)
Arvin M Gouw, Katherine Margulis, Natalie S Liu, Sudha J Raman, Anthony Mancuso, Georgia G Toal, Ling Tong, Adriane Mosley, Annie L Hsieh, Delaney K Sullivan, Zachary E Stine, Brian J Altman, Almut Schulze, Chi V Dang, Richard N Zare, Dean W Felsher
ABSTRACT
Lipid metabolism is frequently perturbed in cancers, but the underlying mechanism is unclear. We present comprehensive evidence that oncogene MYC, in collaboration with transcription factor sterol-regulated element-binding protein (SREBP1), regulates lipogenesis to promote tumorigenesis. We used human and mouse tumor-derived cell lines, tumor xenografts, and four conditional transgenic mouse models of MYC-induced tumors to show that MYC regulates lipogenesis genes, enzymes, and metabolites. We found that MYC induces SREBP1, and they collaborate to activate fatty acid (FA) synthesis and drive FA chain elongation from glucose and glutamine. Further, by employing desorption electrospray ionization mass spectrometry imaging (DESI-MSI), we observed in vivo lipidomic changes upon MYC induction across different cancers, for example, a global increase in glycerophosphoglycerols. After inhibition of FA synthesis, tumorigenesis was blocked, and tumors regressed in both xenograft and primary transgenic mouse models, revealing the vulnerability of MYC-induced tumors to the inhibition of lipogenesis.
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Cerulenin, ≥98% (HPLC), from Cephalosporium caerulens
Sigma-Aldrich
IgG from mouse serum, reagent grade, ≥95% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
IgG from rabbit serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
D-Lysine, ≥98% (HPLC)
Sigma-Aldrich
D-Glucose-1-13C, 99 atom % 13C
Sigma-Aldrich
TOFA, ≥98% (HPLC)
Supelco
Rotenone, PESTANAL®, analytical standard
Sigma-Aldrich
Anti-Fatty Acid Synthase antibody produced in rabbit, 1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Rotenone, ≥95%