  • Search Results
  • Genomic environment influences the dynamics of the tirant LTR retrotransposon in Drosophila.
Genomic environment influences the dynamics of the tirant LTR retrotransposon in Drosophila.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2009-01-15)
Marie Fablet, Emmanuelle Lerat, Rita Rebollo, Béatrice Horard, Nelly Burlet, Sonia Martinez, Emilie Brasset, Eric Gilson, Chantal Vaury, Cristina Vieira
ABSTRACT
Combining genome sequence analysis and functional analysis, we show that some full-length copies of tirant are present in heterochromatic regions in Drosophila simulans and that when tested in vitro, these copies have a functional promoter. However, when inserted in heterochromatic regions, tirant copies are inactive in vivo, and only transcription of euchromatic copies can be detected. Thus, our data indicate that the localization of the element is a hallmark of its activity in vivo and raise the question of genomic invasions by transposable elements and the importance of their genomic integration sites.
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit
Sigma-Aldrich
IgG from rabbit serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder