Merck
Cysteine Toxicity Drives Age-Related Mitochondrial Decline by Altering Iron Homeostasis.
Cell (2020-01-25)
Casey E Hughes, Troy K Coody, Mi-Young Jeong, Jordan A Berg, Dennis R Winge, Adam L Hughes
RESUMEN
Mitochondria and lysosomes are functionally linked, and their interdependent decline is a hallmark of aging and disease. Despite the long-standing connection between these organelles, the function(s) of lysosomes required to sustain mitochondrial health remains unclear. Here, working in yeast, we show that the lysosome-like vacuole maintains mitochondrial respiration by spatially compartmentalizing amino acids. Defects in vacuole function result in a breakdown in intracellular amino acid homeostasis, which drives age-related mitochondrial decline. Among amino acids, we find that cysteine is most toxic for mitochondria and show that elevated non-vacuolar cysteine impairs mitochondrial respiration by limiting intracellular iron availability through an oxidant-based mechanism. Cysteine depletion or iron supplementation restores mitochondrial health in vacuole-impaired cells and prevents mitochondrial decline during aging. These results demonstrate that cysteine toxicity is a major driver of age-related mitochondrial deterioration and identify vacuolar amino acid compartmentation as a cellular strategy to minimize amino acid toxicity.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
S-Methyl-L-cysteine, substrate for methionine sulfoxide reductase A
Sigma-Aldrich
Lys-Lys-Lys-Lys, ≥95% (TLC)
Supelco
Paraquat dichloride hydrate, PESTANAL®, analytical standard
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Methyl viologen dichloride hydrate, 98%
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Iron(III) chloride, reagent grade, 97%
Sigma-Aldrich
L-Homocysteine, ≥98.0% (NT)
Sigma-Aldrich
OxyBlot Protein Oxidation Detection Kit, The OxyBlot Protein Oxidation Detection Kit provides the reagents to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidation.
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Lyticase from Arthrobacter luteus, lyophilized powder, ≥200 units/mg solid
Sigma-Aldrich
Anti-CP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)