• Inicio
  • Resultados de la búsqueda
  • Biotransformation and tissue distribution of protopine and allocryptopine and effects of Plume Poppy Total Alkaloid on liver drug-metabolizing enzymes.
Biotransformation and tissue distribution of protopine and allocryptopine and effects of Plume Poppy Total Alkaloid on liver drug-metabolizing enzymes.
Scientific reports (2018-01-13)
Ya-Jun Huang, Pi Cheng, Zhuo-Yi Zhang, Shi-Jie Tian, Zhi-Liang Sun, Jian-Guo Zeng, Zhao-Ying Liu
RESUMEN
In this study, the biotransformation in the plasma, urine and feces of rats following oral administration of protopine (PRO) and allocryptopine (ALL)were explored using HPLC-QqTOF MS. An HPLC-MS/MS method for the determination of tissues was developed and applied to the tissue distribution study in rats following intragastric administration of Plume Poppy Total Alkaloid for 3 weeks. A total of ten PRO metabolites and ten ALL metabolites were characterized in rats in vivo. Among these metabolites, six PRO metabolites and five ALL metabolites were reported for the first time. The predicated metabolic pathways including ring cleavage, demethylation following ring cleavage, and glucuronidation were proposed. The low-concentration residue of PRO and ALL in various tissues was detected at 24 h and 48 h after dosing, which indicated that both compounds could be widely distributed in tissues and exist as low levels of residue. The activities of erythromycin N-demethylase, aminopyrine N-demethylase and NAD (P)H quinone oxidoreductase in female rats can be induced post-dose, but these activities were inhibited in male rats. The proposed biotransformation and residues of PRO and ALL and their effects on enzymes may provide a basis for clarifying the metabolism and interpreting pharmacokinetics.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Dihydrokainic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Protopine hydrochloride, ≥98%, solid