  • Search Results
  • Newcastle Disease Virus (NDV) Oncolytic Activity in Human Glioma Tumors Is Dependent on CDKN2A-Type I IFN Gene Cluster Codeletion.
Newcastle Disease Virus (NDV) Oncolytic Activity in Human Glioma Tumors Is Dependent on CDKN2A-Type I IFN Gene Cluster Codeletion.
Cells (2020-06-11)
Noemi García-Romero, Irina Palacín-Aliana, Susana Esteban-Rubio, Rodrigo Madurga, Sergio Rius-Rocabert, Josefa Carrión-Navarro, Jesús Presa, Sara Cuadrado-Castano, Pilar Sánchez-Gómez, Adolfo García-Sastre, Estanislao Nistal-Villan, Angel Ayuso-Sacido
ABSTRACT
Glioblastoma (GBM) is the most aggressive and frequent primary brain tumor in adults with a median overall survival of 15 months. Tumor recurrence and poor prognosis are related to cancer stem cells (CSCs), which drive resistance to therapies. A common characteristic in GBM is CDKN2A gene loss, located close to the cluster of type I IFN genes at Ch9p21. Newcastle disease virus (NDV) is an avian paramyxovirus with oncolytic and immunostimulatory properties that has been proposed for the treatment of GBM. We have analyzed the CDKN2A-IFN I gene cluster in 1018 glioma tumors and evaluated the NDV oncolytic effect in six GBM CSCs ex vivo and in a mouse model. Our results indicate that more than 50% of GBM patients have some IFN deletion. Moreover, GBM susceptibility to NDV is dependent on the loss of the type I IFN. Infection of GBM with an NDV-expressing influenza virus NS1 protein can overcome the resistance to oncolysis by NDV of type I-competent cells. These results highlight the potential of using NDV vectors in antitumor therapies.
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine sodium salt, γ-irradiated, powder, suitable for, suitable for cell culture
Sigma-Aldrich
hBFGF, FGF-Basic, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture