Characterisation of the biochemical and cellular roles of native and pathogenic amelogenesis imperfecta mutants of FAM83H.
Cellular signalling (2020-04-15)
Theresa Tachie-Menson, Ana Gázquez-Gutiérrez, Luke J Fulcher, Thomas J Macartney, Nicola T Wood, Joby Varghese, Robert Gourlay, Renata F Soares, Gopal P Sapkota
RESUMEN
The majority of mutations identified in patients with amelogenesis imperfecta have been mapped to FAM83H. As FAM83H expression is not limited to the enamel, how FAM83H contributes to amelogenesis is still largely unknown. We previously reported that members of the FAM83 family of proteins interact with and regulate the subcellular distribution of the promiscuous serine-threonine protein kinase CK1 family, through their shared N-terminal DUF1669 domains. FAM83H co-localises with CK1 isoforms to speckle-like structures in both the cytoplasm and nucleus. In this report, we show FAM83H, unlike other FAM83 proteins, interacts and colocalises with NCK1/2 tyrosine kinase adaptor proteins. This interaction is mediated by proline-rich motifs within the C-terminus of FAM83H, specifically interacting with the second and third SH3 domains of NCK1/2. Moreover, FAM83H pathogenic AI mutant proteins, which trigger C-terminal truncations of FAM83H, retain their interactions with CK1 isoforms but lose interaction with NCK1/2. These AI mutant FAM83H proteins acquire a nuclear localisation, and recruit CK1 isoforms to the nucleus where CK1 retains its kinase activity. As understanding the constituents of the FAM83H-localised speckles may hold the key to unravelling potential substrates of FAM83H-associated CK1 substrates, we employed a TurboID-based proximity labelling approach and uncovered several proteins including Iporin and BAG3 as potential constituents of the speckles.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, 0.2 μm filtered
Sigma-Aldrich
Hexadimethrine bromide, ≥94% (titration)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Gelatin from cold water fish skin, 40-50% in H2O
Millipore
NP-40 Alternative - CAS 9016-45-9 - Calbiochem, A non-ionic surfactant used in the isolation and purification of functional membrane protein complexes and in two-dimensional (2D) gel electrophoresis.
Sigma-Aldrich
Anti-CSNK1E antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Redes sociales
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Investigación. Desarrollo. Producción.
Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos
© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.
Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.