  • Search Results
  • Identification and quantification of N-linked oligosaccharides released from glycoproteins: an inter-laboratory study.
Identification and quantification of N-linked oligosaccharides released from glycoproteins: an inter-laboratory study.
Glycobiology (2008-10-14)
Smita Thobhani, Chun-Ting Yuen, Marc J A Bailey, Christopher Jones
ABSTRACT
As characterization of glycosylation is required for the licensing of recombinant glycoprotein therapeutics, technique comparability must be assessed. Eleven UK laboratories (seven industrial, two regulatory or government, two academic) participated in an inter-laboratory study to analyze N-glycans present in four mixtures prepared by PNGase F cleavage of commercial glycoproteins: human alpha1-acid glycoprotein (H alpha1), bovine alpha1-acid glycoprotein (B alpha1), bovine pancreatic ribonuclease B (RNaseB), and human serum immunoglobulin G (hIgG). Participants applied their routine glycan mapping methodology using predominantly chromatography and mass spectrometry to identify and quantify components. Data interpretation focused on the relative amounts of different glycan structures present, the degree of sialylation, antennary and the galactosylation profiles, fucosylation and bisecting GlcNAc content, and the number of glycan components identified. All laboratories found high levels of sialylation for H alpha1 and B alpha1 (Z-numbers 271 +/- 24 and 224 +/- 18, respectively), but varying ratios of di-, tri-, and tetra-antennary chains. The Z-score for hIgG glycans had high variability as values obtained from mass spectrometric and chromatographic methods clustered separately. The proportion of the major penta-mannosyl chain from RNaseB was between 29 and 62%. Proportions of fucosylated and bisected GlcNAc chains from hIgG were between 58 and 96% and 9 and 23%, respectively. Mass spectrometric approaches consistently identified more glycan species, especially when both N-glycolylneuraminic acid (Neu5Gc) and N-acetylneuraminic acid (Neu5Ac) were present. These data highlight the need for well-characterized reference standards to support method validation and regulatory guidance on selection of approaches. Pharmacopoeial specifications must acknowledge method variability.
Sigma-Aldrich
IgG from human serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder