Merck
The mammalian cholesterol synthesis enzyme squalene monooxygenase is proteasomally truncated to a constitutively active form.
The Journal of biological chemistry (2021-05-03)
Hudson W Coates, Isabelle M Capell-Hattam, Andrew J Brown
RESUMEN
Squalene monooxygenase (SM, also known as squalene epoxidase) is a rate-limiting enzyme of cholesterol synthesis that converts squalene to monooxidosqualene and is oncogenic in numerous cancer types. SM is subject to feedback regulation via cholesterol-induced proteasomal degradation, which depends on its lipid-sensing N-terminal regulatory domain. We previously identified an endogenous truncated form of SM with a similar abundance to full-length SM, but whether this truncated form is functional or subject to the same regulatory mechanisms as full-length SM is not known. Here, we show that truncated SM differs from full-length SM in two major ways: it is cholesterol resistant and adopts a peripheral rather than integral association with the endoplasmic reticulum membrane. However, truncated SM retains full SM activity and is therefore constitutively active. Truncation of SM occurs during its endoplasmic reticulum-associated degradation and requires the proteasome, which partially degrades the SM N-terminus and disrupts cholesterol-sensing elements within the regulatory domain. Furthermore, truncation relies on a ubiquitin signal that is distinct from that required for cholesterol-induced degradation. Using mutagenesis, we demonstrate that partial proteasomal degradation of SM depends on both an intrinsically disordered region near the truncation site and the stability of the adjacent catalytic domain, which escapes degradation. These findings uncover an additional layer of complexity in the post-translational regulation of cholesterol synthesis and establish SM as the first eukaryotic enzyme found to undergo proteasomal truncation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Mevastatin, ≥98% (HPLC), powder or crystals
Sigma-Aldrich
Calpain Inhibitor I, ≥97% (TLC), powder
Sigma-Aldrich
(±)-Mevalonolactone, ~97% (titration)
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
Ponceau S solution, BioReagent, suitable for electrophoresis, 0.1 % (w/v) in 5% acetic acid
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Bromophenol Blue
Sigma-Aldrich
Ethylene glycol-bis(β-aminoethyl ether)-N,N,N′,N′-tetraacetic acid tetrasodium salt, ≥97%
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Cholesterol-Water Soluble, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)