  • Search Results
  • ADH5-mediated NO bioactivity maintains metabolic homeostasis in brown adipose tissue.
ADH5-mediated NO bioactivity maintains metabolic homeostasis in brown adipose tissue.
Cell reports (2021-11-18)
Sara C Sebag, Zeyuan Zhang, Qingwen Qian, Mark Li, Zhiyong Zhu, Mikako Harata, Wenxian Li, Leonid V Zingman, Limin Liu, Vitor A Lira, Matthew J Potthoff, Alexander Bartelt, Ling Yang
ABSTRACT
Brown adipose tissue (BAT) thermogenic activity is tightly regulated by cellular redox status, but the underlying molecular mechanisms are incompletely understood. Protein S-nitrosylation, the nitric-oxide-mediated cysteine thiol protein modification, plays important roles in cellular redox regulation. Here we show that diet-induced obesity (DIO) and acute cold exposure elevate BAT protein S-nitrosylation, including UCP1. This thermogenic-induced nitric oxide bioactivity is regulated by S-nitrosoglutathione reductase (GSNOR; alcohol dehydrogenase 5 [ADH5]), a denitrosylase that balances the intracellular nitroso-redox status. Loss of ADH5 in BAT impairs cold-induced UCP1-dependent thermogenesis and worsens obesity-associated metabolic dysfunction. Mechanistically, we demonstrate that Adh5 expression is induced by the transcription factor heat shock factor 1 (HSF1), and administration of an HSF1 activator to BAT of DIO mice increases Adh5 expression and significantly improves UCP1-mediated respiration. Together, these data indicate that ADH5 controls BAT nitroso-redox homeostasis to regulate adipose thermogenesis, which may be therapeutically targeted to improve metabolic health.
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine sodium salt, γ-irradiated, powder, suitable for, suitable for cell culture
Sigma-Aldrich
S-Nitroso-N-acetyl-DL-penicillamine, ≥97%, powder
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Roche
Bovine Serum Albumin Fraction V, fatty acid free, from bovine serum
Sigma-Aldrich
CL 316,243 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Oil Red O, certified by the Biological Stain Commission
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Indomethacin - CAS 53-86-1 - Calbiochem, A non-steroidal anti-inflammatory, cell permeable, antipyretic agent.