Roles of activin in tissue repair, fibrosis, and inflammatory disease.
Cytokine & growth factor reviews (2006-02-17)
Sabine Werner, Christian Alzheimer
RESUMEN
Activins are transforming growth factor beta superfamily members, which were originally identified as inducers of follicle-stimulating hormone release from the pituitary. In recent years, however, additional functions of activins have been discovered, including a role in the morphogenesis of various tissues and organs. In addition, they have been identified as novel players in inflammation and repair, and are most likely involved in the pathogenesis of inflammatory and fibrotic human diseases. In this review we highlight recent data regarding the expression and function of activins in tissue repair, fibrosis, and inflammation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Monoclonal Anti-Activin B antibody produced in mouse, clone 146807, purified immunoglobulin
Redes sociales
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Investigación. Desarrollo. Producción.
Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos
© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.
Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.