  • Search Results
  • Effect of salicylate on potassium currents in inner hair cells isolated from guinea-pig cochlea.
Effect of salicylate on potassium currents in inner hair cells isolated from guinea-pig cochlea.
Neuroscience letters (2011-09-08)
Takashi Kimitsuki, Mitsuru Ohashi, Yoshihiro Umeno, Takamasa Yoshida, Noritaka Komune, Teppei Noda, Shizuo Komune
ABSTRACT
Although salicylate is one of the most widely used nonsteroidal anti-inflammatory drugs, it causes moderate hearing loss and tinnitus at high-dose levels. In the present study, salicylate effects on the K currents in inner hair cells were examined. Salicylate reversibly reduced the outward K currents (I(K,f)), but did not affect the inward current (I(K,n)). Salicylate blocked the outward K currents in a concentration-dependent manner according to Hill equation with a half-blocking concentration of 1.66mM, and the Hill coefficient of 1.86.
Sigma-Aldrich
Sodium salicylate, puriss. p.a., ≥99.5% (NT)
Sigma-Aldrich
Sodium salicylate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium salicylate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium salicylate, meets USP testing specifications
Sigma-Aldrich
Sodium salicylate, puriss. p.a., reag. Ph. Eur., 99.5-101.0% (calc. to the dried substance)