• Inicio
  • Resultados de la búsqueda
  • Hydrogen sulfide protects PC12 cells against reactive oxygen species and extracellular signal-regulated kinase 1/2-mediated downregulation of glutamate transporter-1 expression induced by chemical hypoxia.
Hydrogen sulfide protects PC12 cells against reactive oxygen species and extracellular signal-regulated kinase 1/2-mediated downregulation of glutamate transporter-1 expression induced by chemical hypoxia.
International journal of molecular medicine (2012-08-17)
Liangcan Xiao, Aiping Lan, Liqiu Mo, Wenming Xu, Nan Jiang, Fen Hu, Jianqiang Feng, Changran Zhang
RESUMEN
Hypoxia and/or ischemia are implicated in neurodegenerative disorders. In these diseases, hypoxia/ischemia may induce oxidative stress, including production of reactive oxygen species (ROS), which result in a decrease in glutamate transporter expression. Hydrogen sulfide (H2S), as the third gasotransmitter, has neuroprotective effects and potent antioxidant properties. In the present study, we investigated the role of glutamate transporter-1 (GLT-1) in the protection of H2S against chemical hypoxia-induced injury in PC12 cells. We found that cobalt chloride (CoCl2), a chemical hypoxia agent, reduced the expression of GLT-1 in a time-dependent manner. Pretreatment with NaHS (a donor of H2S) reversed the CoCl2-induced downregulation of GLT-1 expression. Pretreatment with DHK (a selective inhibitor of GLT-1) for 30 min prior to NaHS preconditioning significantly inhibited the cytoprotection of H2S against CoCl2-induced injuries, leading to an increase in cytotoxicity and apoptosis as well as to a loss of mitochondrial membrane potential (MMP). In addition, we found that similar to the effect of NaHS, pretreatment with NAC (a ROS scavenger) or U0126 (a MEK1/2 inhibitor) blocked the downregulation of GLT-1 expression induced by CoCl2. Collectively, we demonstrated for the first time that ROS and extracellular signal-regulated kinase 1/2 (ERK1/2)-mediated reduction of GLT-1 expression may be involved in chemical hypoxia-induced neural injury and that H2S attenuates this injury partly by upregulating GLT-1 expression in PC12 cells.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Dihydrokainic acid, ≥98% (HPLC), powder