• Inicio
  • Resultados de la búsqueda
  • Synthesis of isoindole and benzoisoindole derivatives of teicoplanin pseudoaglycon with remarkable antibacterial and antiviral activities.
Synthesis of isoindole and benzoisoindole derivatives of teicoplanin pseudoaglycon with remarkable antibacterial and antiviral activities.
Bioorganic & medicinal chemistry letters (2012-10-27)
Attila Sipos, Zsolt Török, Erzsébet Rőth, Attila Kiss-Szikszai, Gyula Batta, Ilona Bereczki, Zsolt Fejes, Anikó Borbás, Eszter Ostorházi, Ferenc Rozgonyi, Lieve Naesens, Pál Herczegh
RESUMEN
The primary amino function of teicoplanin pseudoaglycon has been transformed into arylthioisoindole or benzoisoindole and glycosylthioisoindole derivatives, in a reaction with o-phthalaldehyde or naphtalene-2,3-dicarbaldehyde and various thiols. All of the obtained semisynthetic antibiotics exhibited potent antibacterial activities against Gram-positive bacteria in the ng per ml concentration range. A few of them showed antiviral activity, in particular against influenza virus.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder (may contain lumps)
Sigma-Aldrich
Phthaldialdehyde Reagent, Solution Complete
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder
Sigma-Aldrich
Phthaldialdehyde, for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Teicoplanin
Sigma-Aldrich
Phthaldialdehyde Reagent, Solution Incomplete