Relationship between increase in astrocytic GLT-1 glutamate transport and late-LTP.
Learning & memory (Cold Spring Harbor, N.Y.) (2012-11-21)
Juan D Pita-Almenar, Shengwei Zou, Costa M Colbert, Arnold Eskin
RESUMEN
Na⁺-dependent high-affinity glutamate transporters have important roles in the maintenance of basal levels of glutamate and clearance of glutamate during synaptic transmission. Interestingly, several studies have shown that basal glutamate transport displays plasticity. Glutamate uptake increases in hippocampal slices during early long-term potentiation (E-LTP) and late long-term potentiation (L-LTP). Four issues were addressed in this research: Which glutamate transporter is responsible for the increase in glutamate uptake during L-LTP? In what cell type in the hippocampus does the increase in glutamate uptake occur? Does a single type of cell contain all the mechanisms to respond to an induction stimulus with a change in glutamate uptake? What role does the increase in glutamate uptake play during L-LTP? We have confirmed that GLT-1 is responsible for the increase in glutamate uptake during L-LTP. Also, we found that astrocytes were responsible for much, if not all, of the increase in glutamate uptake in hippocampal slices during L-LTP. Additionally, we found that cultured astrocytes alone were able to respond to an induction stimulus with an increase in glutamate uptake. Inhibition of basal glutamate uptake did not affect the induction of L-LTP, but inhibition of the increase in glutamate uptake did inhibit both the expression of L-LTP and induction of additional LTP. It seems likely that heightened glutamate transport plays an ongoing role in the ability of hippocampal circuitry to code and store information.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
DL-2-Amino-5-phosphonopentanoic acid, solid
Sigma-Aldrich
DNQX, ≥98% (TLC)
Sigma-Aldrich
Dihydrokainic acid, ≥98% (HPLC), powder