Assessment of frequent litter amendment application on ammonia emission from broilers operations.
Journal of the Air & Waste Management Association (1995) (2013-05-22)
Hong Li, Chongyang Lin, Stephen Collier, William Brown, Susan White-Hansen
RESUMEN
Litter amendments have been used to control the ammonia (NH3) emission from the broiler litter during the brooding period. One of the commercially available litter amendments, sodium bisulfate, was frequently applied on the litter with two different rates on weekly basis in a laboratory setup and with a single rate on biweekly basis under field conditions. Repeated application ofsodium bisulfate led to significant reduction in NH3 emissions from broilers. The magnitude of NH3 emission reduction increases with the application rate of sodium bisulfate. The reduction rates of cumulative emissions with 366 g/wk-m2 (75 lb/wk-1000 ft) rate (from 14% to 64.5%) were higher than the reduction rate of 183 g/wk-m2 (37.5 lb/wk-1000 ft2) rate (from 0% to 55%) from 28 to 61 days of age. The cumulative NH3 emission was reduced by 51.7% with 244 g/2 wk-m2 (50 lb/2 wk-1000 ft2) rate over a three-flockperiod (8-wk average grow-out per flock) under field production conditions. Sodium bisulfate application showed no significant difference on body weight and feed conversion efficiency. However, footpad quality was significantly improved by sodium bisulfate application. Litter pH and ammonia nitrogen level of the litter were decreased by sodium bisulfate application with both rates. Organic and total nitrogen contents in the litter were higher, whereas less nitrogen was emitted as NH3. The laboratory-scale findings of emission reduction by the additives should be considered to be preliminary if the additives are to be applied under commercial production settings. This work demonstrated that frequent litter amendment application can be used to reduce NH3 emissions from broiler houses, with no adverse effect on the animal production performances. The NH3 reduction rates could vary with different application frequencies and rates. Using litter amendment during broiler grow-out to lower NH3 emissions should be applicable to boiler production systems. The results of this study also contribute to the baseline data for improving the national air emissions inventory for livestock and poultry production facilities.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Sodium hydrogen sulfate, technical grade
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Sodium sulfate decahydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Sodium bisulfate monohydrate, ReagentPlus®, 99%
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate decahydrate, reagent grade, 97%
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Sodium sulfate decahydrate, BioUltra, ≥99.0% (calc. on dry substance, T)
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate decahydrate, puriss. p.a., crystallized, ≥99.0% (calc. based on dry substance, T)
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.9%