Biogenic silver nanoparticles with chlorogenic acid as a bioreducing agent.
Journal of nanoscience and nanotechnology (2013-07-26)
Hwa Jung Noh, Hyun-Seok Kim, Sang Hui Jun, Young-Hwa Kang, Seonho Cho, Youmie Park
RESUMEN
We report the synthesis of biogenic silver nanoparticles using chlorogenic acid as a bioreducing agent. Chlorogenic acid is a polyphenol compound abundant in coffee. UV-Vis spectra showed the characteristic surface plasmon resonance band at 415 nm, indicating the successful synthesis of biogenic silver nanoparticles. Spherical and irregular shaped nanoparticles were observed with an average diameter of 19.29 +/- 8.23 nm. The reaction yield from silver ion to silver nanoparticles was observed as 95.43% by using inductively coupled plasma-mass spectrometry. Fourier transform infrared spectra revealed that the -C = O groups of chlorogenic acid may coordinate or complex into silver nanoparticles. Biogenic silver nanoparticles exerted higher antibacterial activity against Gram-negative bacteria than against Gram-positive bacteria. Interestingly, a comparable antibacterial activity to a standard antibiotic was observed against two strains of Pseudomonas aeruginosa (minimum inhibitory concentration of 0.66 microg/mL). The synergistic effect of a combination of silver nanoparticles and chlorogenic acid on antibacterial activity is obvious, leading to approximately 8-fold enhancement in the case of Pseudomonas aeruginosa when compared with chlorogenic acid alone. The present report suggests that a pure compound with a plant origin is capable of being a bioreducing agent for the synthesis of biogenic silver nanoparticles with superior antibacterial activity, opening up many applications in nanomedicine and nanobiotechnology.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Sigma-Aldrich
Silver, conductive paste
Supelco
Activated charcoal, powder
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Silver nanowires, diam. × L 60 nm × 10 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Chlorogenic acid, primary reference standard
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Sigma-Aldrich
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Silver nanowires, diam. × L 115 nm × 20-50 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size