Merck
Measurement of free fatty acids in cerebrospinal fluid from patients with hemorrhagic and ischemic stroke.
Brain research (2003-09-12)
J G Pilitsis, W M Coplin, M H O'Regan, J M Wellwood, F G Diaz, M R Fairfax, D B Michael, J W Phillis
RESUMEN
Free fatty acid (FFA) concentrations in cerebrospinal fluid (CSF) from patients with ischemic and hemorrhagic stroke (n=25) and in contemporary controls (n=73) were examined using HPLC. Concentrations of CSF FFAs from ischemic and hemorrhagic stroke patients obtained within 48 h of the insult were significantly greater than in control patients. Higher concentrations of polyunsaturated fatty acids (PUFAs) in CSF obtained within 48 h of insult were associated with significantly lower (P<0.05) admission Glasgow Coma Scale scores and worse outcome at the time of hospital discharge, using the Glasgow Outcome Scale (P<0.01).
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
Supelco
L-Cysteine, certified reference material, TraceCERT®
Sigma-Aldrich
cis-4,7,10,13,16,19-Docosahexaenoic acid, ≥98%
Sigma-Aldrich
DL-Homocysteine, ≥95% (titration)
Sigma-Aldrich
3-Hydroxyanthranilic acid, 97%
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
Adenosine 5′-diphosphate, ≥95% (HPLC)
SAFC
L-Cysteine