A new regulatory motif in cell-cycle control causing specific inhibition of cyclin D/CDK4.
Nature (1993-12-16)
M Serrano, G J Hannon, D Beach
RESUMEN
The division cycle of eukaryotic cells is regulated by a family of protein kinases known as the cyclin-dependent kinases (CDKs). The sequential activation of individual members of this family and their consequent phosphorylation of critical substrates promotes orderly progression through the cell cycle. The complexes formed by CDK4 and the D-type cyclins have been strongly implicated in the control of cell proliferation during the G1 phase. CDK4 exists, in part, as a multi-protein complex with a D-type cyclin, proliferating cell nuclear antigen and a protein, p21 (refs 7-9). CDK4 associates separately with a protein of M(r) 16K, particularly in cells lacking a functional retinoblastoma protein. Here we report the isolation of a human p16 complementary DNA and demonstrate that p16 binds to CDK4 and inhibits the catalytic activity of the CDK4/cyclin D enzymes. p16 seems to act in a regulatory feedback circuit with CDK4, D-type cyclins and retinoblastoma protein.
Redes sociales
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Investigación. Desarrollo. Producción.
Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos
© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.
Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.