The CDK4-pRB-E2F1 pathway : A new modulator of insulin secretion.
Islets (2010-11-26)
Lluis Fajas, Emilie Blanchet, Jean-Sébastien Annicotte
RESUMEN
Pancreatic β-cells are sensors of circulating glucose levels that control insulin secretion through a finely-tuned process. Under hyperglycemic conditions, glucose enters the cell, generates ATP, leading to a subsequent closure of voltage-dependent ATP channels, membrane depolarization, Ca²(+) entry and exocytosis of insulin vesicles. In pathological conditions, such as during type 2 diabetes (T2D), chronic hyperglycemia will ultimately result in decreased capability of β-cells to secrete sufficient amount of insulin to regulate glycemia. Therefore, understanding of the mechanisms of modulation of insulin secretion could be of interest for the treatment of diabetes. We have demonstrated that a particular cell cycle regulator, E2F1, is involved in pancreatic post-natal growth through its functions in the control of β-cell proliferation. Based on the observation that cell cycle regulators were highly expressed in non-proliferating β-cell, we hypothesized that these proteins could also have a direct role in pancreatic β-cell function. Altogether our data unravel a new function for these factors in the control of insulin secretion and open up new avenues for the treatment of diabetes.
Redes sociales
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Investigación. Desarrollo. Producción.
Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos
© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.
Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.